N-Acetylornithine ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Ac-Orn-OH, (S)-2-Acetamido-5-aminopentanoic acid, Ac-Orn-OH, N-Acetyl-L-ornithine

A675142-5G A675142-1G A675142-250MG A675142-25G
77614-084EA 237.93 USD
77614-084 77686-642 77686-643 77893-359
N-Acetylornithine ≥98%
N-Acetylornithine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₃
MDL Number: MFCD00065115
CAS Number: 6205-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR