2-(Hydroxymethyl)acrylic acid ≥95%

Supplier: Ambeed
Danger

A675059-1G A675059-5G
77573-178EA 251 USD
77573-178 77617-816
2-(Hydroxymethyl)acrylic acid ≥95%
2-(Hydroxymethyl)acrylic acid

Precaución: for Research Use Only

Formula: C₄H₆O₃
MDL Number: MFCD01938626
CAS Number: 4370-80-3
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR