2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone, 2-Amino-5-(4-(2-(5-ethylpyridin-2-yl)ethoxy)benzyl)thiazol-4(5H)-one

A674751-5G A674751-1G
77627-364EA 16.48 USD
77627-364 77878-507
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥95%
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O₂S
MDL Number: MFCD07368627
CAS Number: 105355-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR