Necrostatin-7 ≥98%

Supplier: Ambeed
Warning

A674323-10MG A674323-5MG
77747-371EA 149 USD
77747-371 77747-372
Necrostatin-7 ≥98%
Necrostatin-7

Precaución: for Research Use Only

Formula: C₁₆H₁₀FN₅OS₂
MDL Number: MFCD02334728
CAS Number: 351062-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR