4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methylpyridin-1-ium iodide ≥96%

Supplier: Ambeed
Warning

A674045-100MG A674045-1G A674045-250MG A674045-5G
77901-856EA 51 USD
77901-856 77900-916 77900-813 77901-697
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methylpyridin-1-ium iodide ≥96%
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methylpyridin-1-ium iodide

Precaución: for Research Use Only

Formula: C₁₁H₁₇BINO₂
MDL Number: MFCD26523329
CAS Number: 870556-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR