2-Mercapto-1-phenylethanone ≥97%

Supplier: Ambeed
Danger

A673514-100MG A673514-1G
77891-330EA 78 USD
77891-330 77891-331
2-Mercapto-1-phenylethanone ≥97%
2-Mercapto-1-phenylethanone

Precaución: for Research Use Only

Formula: C₈H₈OS
MDL Number: MFCD00661458
CAS Number: 2462-02-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR