6-Bromo-2-(trifluoromethyl)benzo[d]oxazole ≥95%

Supplier: Ambeed
Warning

A673165-1G A673165-5G
77598-690EA 261.38 USD
77598-690 77749-387
6-Bromo-2-(trifluoromethyl)benzo[d]oxazole ≥95%
6-Bromo-2-(trifluoromethyl)benzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO
MDL Number: MFCD11111758
CAS Number: 1260834-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR