4-(1H-Pyrrol-1-yl)benzenesulfonamide ≥97%

Supplier: Ambeed
Warning

A673023-250MG A673023-1G
77747-366EA 154 USD
77747-366 77747-365
4-(1H-Pyrrol-1-yl)benzenesulfonamide ≥97%
4-(1H-Pyrrol-1-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
MDL Number: MFCD01762129
CAS Number: 5438-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR