(4-(3-Fluorophenoxy)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A671764-100MG A671764-1G
77884-563EA 20 USD
77884-563 77884-564
(4-(3-Fluorophenoxy)phenyl)boronic acid ≥98%
(4-(3-Fluorophenoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₃
MDL Number: MFCD14636485
CAS Number: 1029438-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR