Benzyl acetoacetate ≥98%

Supplier: Ambeed
Warning

A671709-100G A671709-25G
77627-348EA 66.95 USD
77627-348 77627-350
Benzyl acetoacetate ≥98%
Benzyl acetoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD00008785
CAS Number: 5396-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR