3-((3S,4R)-3-Methyl-6-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1,6-diazaspiro[3.4]octan-1-yl)-3-oxopropanenitrile ≥99%

Supplier: Ambeed
Warning

A671241-10MG A671241-1MG A671241-5MG A671241-50MG A671241-100MG
77617-804EA 370.8 USD
77617-804 77617-806 77617-808 77749-382 77749-381
3-((3S,4R)-3-Methyl-6-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1,6-diazaspiro[3.4]octan-1-yl)-3-oxopropanenitrile ≥99%
3-((3S,4R)-3-Methyl-6-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1,6-diazaspiro[3.4]octan-1-yl)-3-oxopropanenitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₆O
MDL Number: MFCD31630732
CAS Number: 1263774-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR