5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride ≥98%

Supplier: Ambeed
Warning

A670855-5G A670855-250MG
77627-344EA 528.39 USD
77627-344 77878-503
5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride ≥98%
5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride

Precaución: for Research Use Only

Formula: C₄₈H₃₆ClFeN₄O₄
MDL Number: MFCD00012405
CAS Number: 36995-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR