GSK 1016790A ≥98%

Supplier: Ambeed
Warning

Synonyms: GSK1016790A, N-((S)-1-(4-((S)-2-(2,4-dichlorophenylsulphonamido)-3-hydroxypropanoyl)piperazin-1-yl)-4-methyl-1-oxopentan-2-yl)benzo[b]thiophene-2-carboxamide

A670815-10MG A670815-1MG A670815-5MG
77617-796EA 66.95 USD
77617-796 77617-798 77617-800
GSK 1016790A ≥98%
GSK 1016790A

Precaución: for Research Use Only

Formula: C₂₈H₃₂Cl₂N₄O₆S₂
MDL Number: MFCD12912413
CAS Number: 942206-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR