L(-)-Mannose ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose

A670095-1G A670095-100MG A670095-5G
77602-702EA 144.53 USD
77602-702 77669-826 77669-828
L(-)-Mannose ≥98%
L(-)-Mannose

Precaución: for Research Use Only

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00136021
CAS Number: 10030-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR