SU-6668 ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4-Dimethyl-5-((2-oxoindolin-3-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid, Orantinib

A670041-10MG A670041-1MG A670041-5MG
77614-066EA 74.16 USD
77614-066 77614-068 77614-070
SU-6668 ≥98%
SU-6668

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₃
MW: 310.4 g/mol
MDL Number: MFCD09743433
CAS Number: 252916-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR