4-Chloro-3,5-difluorobenzaldehyde ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A669703-1G A669703-5G
77684-574EA 50 USD
77684-574 77891-325
4-Chloro-3,5-difluorobenzaldehyde ≥98%, Ambeed.Inc
4-Chloro-3,5-difluorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃ClF₂O
MDL Number: MFCD11553555
CAS Number: 1160573-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR