(R)-N-(2-(4'-(2-(Methylsulfonamido)ethyl)-[1,1'-biphenyl]-4-yl)propyl)propane-2-sulfonamide 98+%

Supplier: Ambeed
Warning

Synonyms: Mibampator

A669309-10MG A669309-1MG A669309-5MG
77617-784EA 114.33 USD
77617-784 77617-786 77617-788
(R)-N-(2-(4'-(2-(Methylsulfonamido)ethyl)-[1,1'-biphenyl]-4-yl)propyl)propane-2-sulfonamide 98+%
(R)-N-(2-(4'-(2-(Methylsulfonamido)ethyl)-[1,1'-biphenyl]-4-yl)propyl)propane-2-sulfonamide

Precaución: for Research Use Only

Formula: C₂₁H₃₀N₂O₄S₂
MDL Number: MFCD18251566
CAS Number: 375345-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR