6,6-Dimethyl-3-(5-methyl-3-oxo-2-(4-(trifluoromethoxy)phenyl)-2,3-dihydro-1H-pyrazol-4-yl)-1-phenyl-3-(trifluoromethyl)-6,7-dihydro-1H-indole-2,4(3H,5H)-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: ELOVL6-IN-2

A668331-10MG A668331-5MG
77684-570EA 90 USD
77684-570 77684-571
6,6-Dimethyl-3-(5-methyl-3-oxo-2-(4-(trifluoromethoxy)phenyl)-2,3-dihydro-1H-pyrazol-4-yl)-1-phenyl-3-(trifluoromethyl)-6,7-dihydro-1H-indole-2,4(3H,5H)-dione ≥98%
6,6-Dimethyl-3-(5-methyl-3-oxo-2-(4-(trifluoromethoxy)phenyl)-2,3-dihydro-1H-pyrazol-4-yl)-1-phenyl-3-(trifluoromethyl)-6,7-dihydro-1H-indole-2,4(3H,5H)-dione

Precaución: For research use only.

Formula: C₂₈H₂₃F₆N₃O₄
MW: 579.49 g/mol
MDL Number: MFCD18782631
CAS Number: 1067647-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR