1,1,2,2-Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethene ≥95%

Supplier: Ambeed
Warning

A667747-1G A667747-250MG A667747-5G
77617-774EA 89.61 USD
77617-774 77749-372 77749-373
1,1,2,2-Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethene ≥95%
1,1,2,2-Tetrakis(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethene

Precaución: for Research Use Only

Formula: C₅₀H₆₄B₄O₈
MW: 836.2824 g/mol
CAS Number: 1660996-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR