Streptozocin 98% ≥85% α-anomer basis

Supplier: Ambeed
Danger

A667625-100MG A667625-250MG
77602-696EA 23.58 USD
77602-696 77602-698
Streptozocin 98% ≥85% α-anomer basis
Streptozocin

Precaución: for Research Use Only

Formula: C₈H₁₅N₃O₇
MDL Number: MFCD00006607
CAS Number: 18883-66-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR