3-Chloro-4-formylphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-formylbenzeneboronic acid, (3-Chloro-4-formylphenyl)boronic acid

A667442-250MG A667442-100MG
77673-830EA 45 USD
77673-830 77882-522
3-Chloro-4-formylphenylboronic acid ≥98%
3-Chloro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MDL Number: MFCD08274473
CAS Number: 1072952-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR