Methyl 2,4-dihydroxy-6-pentylbenzoate ≥95%

Supplier: Ambeed
Warning

A667154-100G A667154-1G
77617-772EA 328.57 USD
77617-772 77747-355
Methyl 2,4-dihydroxy-6-pentylbenzoate ≥95%
Methyl 2,4-dihydroxy-6-pentylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₄
MDL Number: MFCD06737776
CAS Number: 58016-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR