tert-Butyl 2-methyl-3-oxopyrrolidine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A667135-1G A667135-250MG
77595-014EA 418.2 USD
77595-014 77684-568
tert-Butyl 2-methyl-3-oxopyrrolidine-1-carboxylate ≥97%
tert-Butyl 2-methyl-3-oxopyrrolidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₇NO₃
MW: 199.24 g/mol
MDL Number: MFCD22566674
CAS Number: 1539946-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR