(2E,6Z,8E,10E)-N-(2-Hydroxy-2-methylpropyl)dodeca-2,6,8,10-tetraenamide ≥95%

Supplier: Ambeed
Warning

Synonyms: Hydroxy-α-sanshool

A666937-10MG A666937-5MG
77673-826EA 240 USD
77673-826 77673-828
(2E,6Z,8E,10E)-N-(2-Hydroxy-2-methylpropyl)dodeca-2,6,8,10-tetraenamide ≥95%
(2E,6Z,8E,10E)-N-(2-Hydroxy-2-methylpropyl)dodeca-2,6,8,10-tetraenamide

Precaución: for Research Use Only

Formula: C₁₆H₂₅NO₂
MW: 263.38 g/mol
MDL Number: MFCD17779317
CAS Number: 83883-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR