Trimethyl-β-cyclodextrin ≥98%

Supplier: Ambeed
Warning

Synonyms: Heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin, Trimethyl-β-cyclodextrin

A666606-1G A666606-250MG
77614-054EA 81.37 USD
77614-054 77686-634
Trimethyl-β-cyclodextrin ≥98%
Trimethyl-beta-cyclodextrin

Precaución: for Research Use Only

Formula: C₆₃H₁₁₂O₃₅
MW: 1429.55 g/mol
MDL Number: MFCD00010728
CAS Number: 55216-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR