tert-Butyl 3,9-diazabicyclo[4.2.1]nonane-9-carboxylate ≥97%

Supplier: Ambeed
Warning

A666469-100MG A666469-1G A666469-250MG
77902-026EA 133 USD
77902-026 77901-477 77900-909
tert-Butyl 3,9-diazabicyclo[4.2.1]nonane-9-carboxylate ≥97%
tert-Butyl 3,9-diazabicyclo[4.2.1]nonane-9-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD18785763
CAS Number: 1251015-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR