Polyoxyethylene (20) sorbitan monolaurate Hydroxyl Value 90-110 mg KOH/g

Supplier: Ambeed
Warning

A666205-100G A666205-500G A666205-1000G
77573-146EA 9 USD
77573-146 77669-814 77669-812
Polyoxyethylene (20) sorbitan monolaurate Hydroxyl Value 90-110 mg KOH/g
Polyoxyethylene (20) sorbitan monolaurate

Precaución: for Research Use Only

MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR