3-Methoxy-5-(methoxycarbonyl)phenylboronic acid 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (3-Methoxy-5-(methoxycarbonyl)phenyl)boronic acid

A665182-250MG A665182-50MG A665182-1G A665182-100MG
77684-566EA 81 USD
77684-566 77684-567 77893-346 77893-345
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid 98+%, Ambeed.Inc
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid

Precaución: For research use only.

Formula: C₉H₁₁BO₅
MW: 209.992 g/mol
MDL Number: MFCD13191801
CAS Number: 1048330-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR