N-Lauroylsarcosine ≥97%

Supplier: Ambeed
Warning

Synonyms: N-Lauroyl-Sar-OH, 2-(N-Methyldodecanamido)acetic acid, N-Dodecanoylsarcosine

A665050-100G A665050-25G
77602-692EA 64.58 USD
77602-692 77602-694
N-Lauroylsarcosine ≥97%
N-Lauroylsarcosine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH
MW: 271.4 g/mol
MDL Number: MFCD00021749
CAS Number: 97-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR