2-(4-Bromophenyl)propan-2-amine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A664905-5G A664905-1G A664905-250MG
77684-564EA 224 USD
77684-564 77893-343 77893-344
2-(4-Bromophenyl)propan-2-amine ≥98%, Ambeed.Inc
2-(4-Bromophenyl)propan-2-amine

Precaución: For research use only.

Formula: C₉H₁₂BrN
MDL Number: MFCD06797600
CAS Number: 17797-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR