Ethyl 5-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A664831-5G A664831-10G
77617-770EA 100.94 USD
77617-770 77749-369
Ethyl 5-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥98%
Ethyl 5-bromoimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.09 g/mol
MDL Number: MFCD06796255
CAS Number: 135995-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR