4-Chloro-3-(trifluoromethoxy)phenylacetic acid ≥98%

Supplier: Ambeed
Warning

A664782-1G A664782-250MG A664782-100MG
77884-549EA 46 USD
77884-549 77884-550 77884-548
4-Chloro-3-(trifluoromethoxy)phenylacetic acid ≥98%
4-Chloro-3-(trifluoromethoxy)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₃
MDL Number: MFCD06660251
CAS Number: 886501-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR