Quinazolin-6-ylboronic acid ≥95%

Supplier: Ambeed
Warning

A663465-100MG A663465-1G A663465-250MG
77617-766EA 212.18 USD
77617-766 77617-768 77749-364
Quinazolin-6-ylboronic acid ≥95%
Quinazolin-6-ylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BN₂O₂
MDL Number: MFCD13189456
CAS Number: 2265882-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR