(6aR,11aR)-9-Methoxy-6a,11a-dihydro-6H-benzofuro[3,2-c]chromen-3-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: Medicarpin

A663354-1MG A663354-10MG A663354-5MG
77614-048EA 66.95 USD
77614-048 77686-629 77686-630
(6aR,11aR)-9-Methoxy-6a,11a-dihydro-6H-benzofuro[3,2-c]chromen-3-ol ≥98%
(6aR,11aR)-9-Methoxy-6a,11a-dihydro-6H-benzofuro[3,2-c]chromen-3-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
MDL Number: MFCD00161527
CAS Number: 32383-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR