4-(3,4-Dihydroquinolin-1(2H)-ylsulfonyl)aniline ≥95%

Supplier: Ambeed
Warning

A662088-1G A662088-250MG
77891-310EA 147 USD
77891-310 77891-311
4-(3,4-Dihydroquinolin-1(2H)-ylsulfonyl)aniline ≥95%
4-(3,4-Dihydroquinolin-1(2H)-ylsulfonyl)aniline

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₂S
MDL Number: MFCD03002135
CAS Number: 5455-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR