4-Bromo-1-ethyl-2-methoxybenzene ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A662060-1G A662060-100MG A662060-250MG
77684-558EA 230 USD
77684-558 77893-336 77893-337
4-Bromo-1-ethyl-2-methoxybenzene ≥95%, Ambeed.Inc
4-Bromo-1-ethyl-2-methoxybenzene

Precaución: For research use only.

Formula: C₉H₁₁BrO
MDL Number: MFCD22490703
CAS Number: 876299-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR