tert-butyl N-[(1R)-3-oxocyclohexyl]carbamate ≥98%

Supplier: Ambeed
Warning

A662027-250MG A662027-100MG
77598-678EA 95.33 USD
77598-678 77783-614
tert-butyl N-[(1R)-3-oxocyclohexyl]carbamate ≥98%
tert-butyl N-[(1R)-3-oxocyclohexyl]carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
MDL Number: MFCD30181863
CAS Number: 1383797-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR