5-Bromo-2-(4-fluorophenyl)pyridine 99+% (by HPLC)
Supplier: Ambeed
|
Warning
|
A661365-100MG
A661365-1G
A661365-250MG
77884-539EA
171
USD
77884-539
77884-540
77884-541
5-Bromo-2-(4-fluorophenyl)pyridine 99+% (by HPLC)
5-Bromo-2-(4-fluorophenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₇BrFN |
MDL Number:
MFCD11100950 CAS Number: 463336-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...