(S)-4-Ethyl-4-hydroxy-11-(2-(isopropylamino)ethyl)-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride ≥97%

Supplier: Ambeed
Danger

Synonyms: Belotecan hydrochloride

A661005-100MG A661005-10MG A661005-1MG A661005-250MG A661005-50MG
77602-684EA 328 USD
77602-684 77627-324 77627-326 77627-328 77627-330
(S)-4-Ethyl-4-hydroxy-11-(2-(isopropylamino)ethyl)-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride ≥97%
(S)-4-Ethyl-4-hydroxy-11-(2-(isopropylamino)ethyl)-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₂₈ClN₃O₄
MW: 469.96 g/mol
MDL Number: MFCD07772313
CAS Number: 213819-48-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR