6-Bromo-2-chloro-3-iodopyridine ≥98%

Supplier: Ambeed
Danger

A660422-25G A660422-1G
77882-510EA 112 USD
77882-510 77882-509
6-Bromo-2-chloro-3-iodopyridine ≥98%
6-Bromo-2-chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MDL Number: MFCD11857690
CAS Number: 1138444-17-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR