3-tert-Butoxycarbonylamino-pentanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)pentanoic acid

A660393-250MG A660393-1G
77614-040EA 65.92 USD
77614-040 77673-800
3-tert-Butoxycarbonylamino-pentanoic acid ≥97%
3-tert-Butoxycarbonylamino-pentanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.262 g/mol
MDL Number: MFCD05863633
CAS Number: 557091-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR