2-[2-(Phenylthio)phenyl]acetic acid ≥98%

Supplier: Ambeed
Warning

A660028-1G A660028-100MG A660028-5G
77669-796EA 45 USD
77669-796 77873-061 77873-062
2-[2-(Phenylthio)phenyl]acetic acid ≥98%
2-[2-(Phenylthio)phenyl]acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂S
MDL Number: MFCD01312192
CAS Number: 1527-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR