3,4-Dichloro-α,α,α-trifluorotoluene ≥97%

Supplier: Ambeed
Danger

A659995-100G A659995-25G A659995-500G A659995-5G
77617-760EA 20.6 USD
77617-760 77749-359 77749-360 77749-361
3,4-Dichloro-α,α,α-trifluorotoluene ≥97%
3,4-Dichloro-α,α,α-trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃
MDL Number: MFCD00000555
CAS Number: 328-84-7
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR