2-Carbamoylisonicotinic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A659646-100MG A659646-1G A659646-250MG
77684-550EA 53 USD
77684-550 77684-551 77893-330
2-Carbamoylisonicotinic acid ≥95%, Ambeed.Inc
2-Carbamoylisonicotinic acid

Precaución: For research use only.

Formula: C₇H₆N₂O₃
MDL Number: MFCD16988758
CAS Number: 160601-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR