4-Amino-1,8-naphthalimide ≥95%

Supplier: Ambeed
Warning

A659621-1G A659621-250MG
77749-357EA 298 USD
77749-357 77749-358
4-Amino-1,8-naphthalimide ≥95%
4-Amino-1,8-naphthalimide

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₂
MDL Number: MFCD00006921
CAS Number: 1742-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR