5-(Trifluoromethyl)-1H-1,2,3-triazole ≥95%

Supplier: Ambeed
Warning

A659609-250MG A659609-1G
77573-132EA 351 USD
77573-132 77891-300
5-(Trifluoromethyl)-1H-1,2,3-triazole ≥95%
5-(Trifluoromethyl)-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₂F₃N₃
MDL Number: MFCD27920868
CAS Number: 40964-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR