JC-1 (iodide) ≥99%

Supplier: Ambeed
Warning

A659203-1MG A659203-2MG A659203-5MG A659203-10MG
77617-758EA 48.41 USD
77617-758 77747-347 77747-348 77747-346
JC-1 (iodide) ≥99%
JC-1 (iodide)

Precaución: for Research Use Only

Formula: C₂₅H₂₇Cl₄IN₄
MDL Number: MFCD00188475
CAS Number: 3520-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR