2-(2-Isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

A659086-1G A659086-250MG
77614-034EA 232.78 USD
77614-034 77614-036
2-(2-Isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid ≥95%
2-(2-Isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₅O₄
MW: 279.25 g/mol
MDL Number: MFCD23703138
CAS Number: 172405-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR