Methyl 4-aminobicyclo[2.2.1]heptane-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A659062-1G A659062-100MG A659062-250MG A659062-50MG
77573-128EA 457 USD
77573-128 77893-327 77893-328 77893-329
Methyl 4-aminobicyclo[2.2.1]heptane-1-carboxylate ≥97%
Methyl 4-aminobicyclo[2.2.1]heptane-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂
MDL Number: MFCD23378922
CAS Number: 1252672-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR