Methyl 2-aminobenzo[d]thiazole-5-carboxylate ≥97%
Supplier: Ambeed
|
Warning
|
A658336-10G
A658336-1G
A658336-250MG
77901-944EA
1518
USD
77901-944
77901-326
77901-262
Methyl 2-aminobenzo[d]thiazole-5-carboxylate ≥97%
Methyl 2-aminobenzo[d]thiazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂S |
MDL Number:
MFCD09263763 CAS Number: 209459-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...